missing translation for 'onlineSavingsMsg'
Learn More

p-Terphenyl (purified by sublimation) 99.5+%, TCI America™

Catalog No. p-7137665
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
T32635G 5 g
T326325G 25 g
2 options

Catalog No. T32635G

Supplier: TCI America T32635G

Only null left

Chemical Identifiers

CAS 92-94-4
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00003061
InChI Key XJKSTNDFUHDPQJ-UHFFFAOYSA-N
Synonym p-terphenyl, 1,1':4',1-terphenyl, p-diphenylbenzene, p-triphenyl, santowax p, 4-phenylbiphenyl, 4-phenyldiphenyl, para-terphenyl, biphenyl, 4-phenyl, pyrogallol tannin
PubChem CID 7115
ChEBI CHEBI:52242
IUPAC Name 1,4-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3

Specifications

Melting Point 214°C
Color White
Boiling Point 389°C
Quantity 5 g
Formula Weight 230.31
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p-Terphenyl (purified by sublimation)
EINECSNumber (4)-0017
RTECSNumber WZ6475000
TSCA Yes