missing translation for 'onlineSavingsMsg'
Learn More

p,p'-Ditolylamine 98.0+%, TCI America™

Catalog No. p-7126517
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D09505G 5 g
D095025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D09505G Supplier TCI America Supplier No. D09505G
Only null left

Chemical Identifiers

CAS 620-93-9
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00059317
InChI Key RHPVVNRNAHRJOQ-UHFFFAOYSA-N
Synonym 4,4'-dimethyldiphenylamine, di-p-tolylamine, p,p'-ditolylamine, 4-methyl-n-4-methylphenyl aniline, benzenamine, 4-methyl-n-4-methylphenyl, bis 4-methylphenyl amine, 4,4-dimethyldiphenylamine, dip-tolylamine, di-p-tolylamin, di-p-tolyl-amine
PubChem CID 69293
IUPAC Name 4-methyl-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)NC2=CC=C(C=C2)C

Specifications

Melting Point 79°C
Boiling Point 330°C
Quantity 5 g
Formula Weight 197.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material p,p′-Ditolylamine
EINECSNumber (4)-1686
TSCA Yes