accessibility menu, dialog, popup

UserName

CAS RN 620-93-9

IUPAC Name: 4-methyl-N-(4-methylphenyl)aniline
Synonyms: 4-methyl-N-(4-methylphenyl)aniline
Molecular Weight (g/mol): 197.28
Molecular Formula: C14H15N
InChi Key: RHPVVNRNAHRJOQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=CC=C(C)C=C2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)