missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Naphtholbenzein TS, 0.25% (w/v) in Glacial Acetic Acid, Ricca Chemical
$207.40 - $372.17
Chemical Identifiers
| CAS | 145-50-6 |
|---|---|
| Molecular Formula | C27H18O2 |
| Molecular Weight (g/mol) | 374.439 |
| InChI Key | VDDWRTZCUJCDJM-PNHLSOANSA-N |
| PubChem CID | 5941340 |
| IUPAC Name | (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one |
| SMILES | C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O |
Chemical Identifiers
| 145-50-6 | |
| 374.439 | |
| 5941340 | |
| C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O |
| C27H18O2 | |
| VDDWRTZCUJCDJM-PNHLSOANSA-N | |
| (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one |
Specifications
| Red | |
| 98.0,0.23 | |
| C27H18O2 | |
| VDDWRTZCUJCDJM-PNHLSOANSA-N | |
| (4Z)-4-[(4-hydroxynaphthalen-1-yl)-phenylmethylidene]naphthalen-1-one | |
| 5941340 | |
| Laboratory | |
| Glacial Acetic Acid | |
| 120 mL | |
| 0.25% (w/v) |
| 145-50-6 , 64-19-7 | |
| 100.0,0.24 | |
| p-naphtholbenzein, 1-naphtholbenzein, alpha-naphtholbenzein, .alpha.-naphtholbenzein, 4-4-hydroxynaphthyl phenylmethylene naphthalen-1-one, naphtholbenzein, 1 4h-naphthalenone, 4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone,4-4-hydroxy-1-naphthalenyl phenylmethylene, 1 4h-naphthalenone, 4-.alpha.-4-hydroxy-1-naphthyl benzylidene | |
| C1=CC=C(C=C1)C(=C2C=CC(=O)C3=CC=CC=C23)C4=CC=C(C5=CC=CC=C54)O | |
| 374.439 | |
| 0.25% | |
| Amber Glass Bottle | |
| <2 | |
| Liquid | |
| p-Naphtholbenzein, Test Solution |