missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Dimethylaminobenzalrhodanine, 0.02% (w/v) in Acetone, Indicator for Argentimetric Titrations, Ricca Chemical
$158.74 - $158.74
Chemical Identifiers
| CAS | 536-17-4 |
|---|---|
| Molecular Formula | C12H12N2OS2 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | JJRVRELEASDUMY-UHFFFAOYSA-N |
| PubChem CID | 2723826 |
| IUPAC Name | 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one |
| SMILES | CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2 |
Chemical Identifiers
| 536-17-4 | |
| Mixture | |
| 2723826 | |
| CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2 |
| C12H12N2OS2 | |
| JJRVRELEASDUMY-UHFFFAOYSA-N | |
| 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one |
Specifications
| 536-17-4 , 67-64-1 | |
| 100.0,0.03 | |
| C12H12N2OS2 | |
| CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2 | |
| Mixture | |
| Indicator | |
| Acetone | |
| 120 mL | |
| p-Dimethylaminobenzalrhodanine, Indicator For Argentimetric Titrations |
| 98.0,0.02 | |
| 0.02% (w/v) | |
| JJRVRELEASDUMY-UHFFFAOYSA-N | |
| 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one | |
| 2723826 | |
| Amber Glass Bottle | |
| Orange | |
| Liquid |