missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Dimethylaminobenzalrhodanine, 0.02% (w/v) in Acetone, Indicator for Argentimetric Titrations, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0, 0.02 |
| CAS Max % | 100.0, 0.03 |
| Concentration | 0.02% (w/v) |
| Packaging | Amber Glass Bottle |
| Solvent or Matrix | Acetone |
| Color | Orange |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Grade | Indicator |
| Chemical Name or Material | p-Dimethylaminobenzalrhodanine, Indicator For Argentimetric Titrations |