missing translation for 'onlineSavingsMsg'
Learn More

p-Dimethylaminobenzaldehyde Solution, Indicator for Hydrazine, Ricca Chemical

Catalog No. p-7239348
Change view
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
2555250 250 mL Amber Glass Bottle
255516 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2555250 Supplier Ricca Chemical Company Supplier No. RI25558
Only null left

Chemical Identifiers

CAS 67-56-1 , 7732-18-5 , 7647-01-0 , 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) Mixture
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

MDL Number MFCD00003381
Grade Laboratory
Packaging Amber Glass Bottle
Color Pale Yellow
Quantity 250 mL
Chemical Name or Material p-Dimethylaminobenzaldehyde solution