Learn More
p-Dimethylaminobenzaldehyde Solution, For Hydrazine, Nickel, Certified, 1.8%, LabChem™
$230.79 - $230.79
Chemical Identifiers
| CAS | 100-10-7 |
|---|---|
| Molecular Formula | CH4O |
| Molecular Weight (g/mol) | 32.04 |
| MDL Number | MFCD00004595 |
| InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
| Synonym | 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal |
| IUPAC Name | methanol |
| SMILES | CO |
Chemical Identifiers
| 100-10-7 | |
| 32.04 | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol |
| CH4O | |
| MFCD00004595 | |
| 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal | |
| CO |
Specifications
| Yellow | |
| 1.79,89.28,8.93 | |
| C9H11NO | |
| UN2924 | |
| Soluble in water | |
| CO | |
| 32.04 | |
| 1.8% | |
| Passes Test | |
| Passes Test | |
| 500 mL | |
| p-Dimethylaminobenzaldehyde Solution, For Hydrazine, Nickel |
| 100-10-7 , 67-56-1 , 7647-01-0 | |
| CH4O | |
| MFCD00004595 | |
| 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal | |
| OKKJLVBELUTLKV-UHFFFAOYSA-N | |
| methanol | |
| 149.19 | |
| Certified | |
| Amber Glass | |
| Hydrogen chloride; Carbon monoxide; Carbon dioxide; Gaseous Ammonia | |
| Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs (central nervous system, optic nerve).
GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature