missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene 95.0+%, TCI America™

Supplier:  TCI America C0513500ML

 View more versions of this product

Catalog No. C0513500ML


Only null left

Description

Description

Specifications

Specifications

p-Cymene
-68°C
176°C
MFCD00008893
2046
HFPZCAJZSCWRBC-UHFFFAOYSA-N
1-methyl-4-propan-2-ylbenzene
7463
134.22
Liquid
99-87-6
Colorless
C10H14
500 mL
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
CC1=CC=C(C=C1)C(C)C
134.222
CHEBI:28768
≥95.0% (GC)

Chemical Identifiers

99-87-6
134.222
HFPZCAJZSCWRBC-UHFFFAOYSA-N
7463
1-methyl-4-propan-2-ylbenzene
C10H14
MFCD00008893
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
CHEBI:28768
CC1=CC=C(C=C1)C(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0012&(3)-0015

RTECSNumber : GZ5950000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions