missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene 95.0+%, TCI America™

Catalog No. p-7124481
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C051325ML 25 mL
C0513500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C051325ML Supplier TCI America Supplier No. C051325ML
Only null left

Chemical Identifiers

CAS 99-87-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008893
InChI Key HFPZCAJZSCWRBC-UHFFFAOYSA-N
Synonym p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
PubChem CID 7463
ChEBI CHEBI:28768
IUPAC Name 1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C

Specifications

Melting Point -68°C
Color Colorless
Boiling Point 176°C
Quantity 25 mL
UN Number 2046
Formula Weight 134.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Cymene
EINECSNumber (3)-0012&(3)-0015
RTECSNumber GZ5950000
TSCA Yes