accessibility menu, dialog, popup

UserName

H3CCH3CH3

CAS RN 99-87-6

IUPAC Name: 1-methyl-4-(propan-2-yl)benzene
Synonyms: cymene 对伞花烃 P-isopropyltoluene camphogen P-cymol P-cimene P-methylcumene P-I-propyltoluene 4-isopropylbenzyl radical isopropyltoluene
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: HFPZCAJZSCWRBC-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C)C=C1
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (4)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (4)
  • (5)
  • (4)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)