missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene, 99+%

CAS: 99-87-6 | C10H14 | 134.22 g/mol

Supplier:  Thermo Scientific Chemicals 111760010

 View more versions of this product

Catalog No. AC111760010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Cymene
-68.0°C
176.0°C to 178.0°C
Authentic
Glass bottle
1.4890 to 1.4910
MFCD00008893
05, II, 322
15, 2760
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,chloroform,acetone,,benzene and ccl4
CC1=CC=C(C=C1)C(C)C
134.22
CHEBI:28768
99+%
99-87-6
0.8540g/mL
47°C
99% min. (GC)
C10H14
CH3C6H4CH(CH3)2
1 L
0.854
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
HFPZCAJZSCWRBC-UHFFFAOYSA-N
1-methyl-4-propan-2-ylbenzene
7463
134.22
Liquid

Chemical Identifiers

99-87-6
134.22
HFPZCAJZSCWRBC-UHFFFAOYSA-N
7463
1-methyl-4-propan-2-ylbenzene
C10H14
MFCD00008893
p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
CHEBI:28768
CC1=CC=C(C=C1)C(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/attentio

GHS Signal Word: Danger

EINECSNumber : 202-796-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only