missing translation for 'onlineSavingsMsg'
Learn More

p-Cymene, 99+%

Catalog No. p-3778096
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111762500 250 mL Glass bottle
AC111760010 1 L Glass bottle
AC111760025 2.5 L Glass bottle
AC111760100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC111762500 Supplier Thermo Scientific Chemicals Supplier No. 111762500
Only null left

CAS: 99-87-6 | C10H14 | 134.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-87-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008893
InChI Key HFPZCAJZSCWRBC-UHFFFAOYSA-N
Synonym p-cymene, 4-isopropyltoluene, p-isopropyltoluene, dolcymene, p-cymol, paracymene, cymene, para-cymene, 1-isopropyl-4-methylbenzene, camphogen
PubChem CID 7463
ChEBI CHEBI:28768
IUPAC Name 1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C

Specifications

Melting Point -68.0°C
Density 0.8540g/mL
Boiling Point 176.0°C to 178.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910
Linear Formula CH3C6H4CH(CH3)2
Quantity 250 mL
Beilstein 05, II, 322
Specific Gravity 0.854
Merck Index 15, 2760
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,chloroform,acetone,,benzene and ccl4
Formula Weight 134.22
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material p-Cymene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/attentio
EINECSNumber 202-796-7

RUO – Research Use Only