missing translation for 'onlineSavingsMsg'
Learn More

p-Cresol, 99%

CAS: 106-44-5 | C7H8O | 108.14 g/mol

Supplier:  Thermo Scientific Chemicals A135310I

 View more versions of this product

Catalog No. AAA135310I


Only null left
Explore available promotions

Description

Description

p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.

Solubility
Soluble in ethanol, diethyl ether and acetone. Slightly soluble in water.

Notes
Air and light sensitive. Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents and bases.
Specifications

Specifications

p-Cresol
32°C to 34°C
201°C to 202°C
Phenol-like
MFCD00002376
UN3455
Light sensitive
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
IWDCLRJOBJJRNH-UHFFFAOYSA-N
4-methylphenol
2879
108.14
106-44-5
1.034
89°C (192°F)
C7H8O
5000 g
1305151
14,2579
Soluble in ethanol,diethyl ether and acetone. Slightly soluble in water.
CC1=CC=C(C=C1)O
108.14
CHEBI:17847
99%

Chemical Identifiers

106-44-5
108.14
IWDCLRJOBJJRNH-UHFFFAOYSA-N
2879
4-methylphenol
C7H8O
MFCD00002376
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene
CHEBI:17847
CC1=CC=C(C=C1)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H311-H314-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CRESOLS, SOLID

EINECSNumber : 203-398-6

RTECSNumber : GO6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only