Learn More
p-Cresol, 99%
CAS: 106-44-5 | C7H8O | 108.14 g/mol
Supplier: Thermo Scientific Chemicals A135310B
Description
p-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Cresol is a starting material in the synthesis of bupranolol which is a non-selective beta blocker. It is also used in the production of antioxidants like butylated hydroxytoluene. Furthermore, it is used as an adhesive and a sealant in chemicals, dyes, intermediates, odor agents, plasticizers, plating agents and surface treating agents. It acts as an antioxidant and a disinfectant in pharmaceuticals.
Solubility
Soluble in ethanol, diethyl ether and acetone. Slightly soluble in water.
Notes
Air and light sensitive. Hygroscopic. Store in cool place. Incompatible with strong oxidizing agents and bases.
Specifications
| p-Cresol | |
| 32°C to 34°C | |
| 201°C to 202°C | |
| Phenol-like | |
| MFCD00002376 | |
| UN3455 | |
| Light sensitive | |
| p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene | |
| IWDCLRJOBJJRNH-UHFFFAOYSA-N | |
| 4-methylphenol | |
| 2879 | |
| 108.14 |
| 106-44-5 | |
| 1.034 | |
| 89°C (192°F) | |
| C7H8O | |
| 1000 g | |
| 1305151 | |
| 14,2579 | |
| Soluble in ethanol,diethyl ether and acetone. Slightly soluble in water. | |
| CC1=CC=C(C=C1)O | |
| 108.14 | |
| CHEBI:17847 | |
| 99% |
Chemical Identifiers
| 106-44-5 | |
| 108.14 | |
| IWDCLRJOBJJRNH-UHFFFAOYSA-N | |
| 2879 | |
| 4-methylphenol |
| C7H8O | |
| MFCD00002376 | |
| p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene | |
| CHEBI:17847 | |
| CC1=CC=C(C=C1)O |
Safety and Handling
GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311-H314-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CRESOLS, SOLID
EINECSNumber : 203-398-6
RTECSNumber : GO6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only