missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine-3-sulfonic Acid 98.0+%, TCI America™

Catalog No. p-7117289
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11775G 5 g
A117725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11775G Supplier TCI America Supplier No. A11775G
Only null left

Chemical Identifiers

CAS 6470-17-3
Molecular Formula C7H9NO4S
Molecular Weight (g/mol) 203.212
MDL Number MFCD00035760
InChI Key JXZGTFLJFKLVAX-UHFFFAOYSA-N
Synonym 4-Aminoanisole-2-sulfonic Acid, 3-Amino-6-methoxybenzenesulfonic Acid, 4-Methoxyaniline-3-sulfonic Acid
PubChem CID 80945
IUPAC Name 5-amino-2-methoxybenzenesulfonic acid
SMILES COC1=C(C=C(C=C1)N)S(=O)(=O)O

Specifications

Color Gray-White
Quantity 5 g
Formula Weight 203.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Anisidine-3-sulfonic Acid
EINECSNumber (3)-2039
TSCA Yes