CAS RN 6470-17-3
IUPAC Name:
5-amino-2-methoxybenzene-1-sulfonic acid
Synonyms:
5-amino-2-methoxybenzenesulfonic acid
Molecular Weight (g/mol):
203.21
Molecular Formula:
C7H9NO4S
InChi Key:
JXZGTFLJFKLVAX-UHFFFAOYSA-N
SMILES:
COC1=CC=C(N)C=C1S(O)(=O)=O
1
–
1
of
1
results
1
p-Anisidine-3-sulfonic Acid 98.0+%, TCI America™
CAS: 6470-17-3 Molecular Formula: C7H9NO4S Molecular Weight (g/mol): 203.212 MDL Number: MFCD00035760 InChI Key: JXZGTFLJFKLVAX-UHFFFAOYSA-N Synonym: 4-Aminoanisole-2-sulfonic Acid, 3-Amino-6-methoxybenzenesulfonic Acid, 4-Methoxyaniline-3-sulfonic Acid PubChem CID: 80945 IUPAC Name: 5-amino-2-methoxybenzenesulfonic acid SMILES: COC1=C(C=C(C=C1)N)S(=O)(=O)O