missing translation for 'onlineSavingsMsg'
Learn More

p-Anisidine-2-sulfonic Acid 97.0+%, TCI America™

Catalog No. p-7117349
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A128525G 25 g
A1285500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A128525G Supplier TCI America Supplier No. A128525G

Chemical Identifiers

CAS 13244-33-2
Molecular Formula C7H9NO4S
Molecular Weight (g/mol) 203.212
MDL Number MFCD00035804
InChI Key KZKGEEGADAWJFS-UHFFFAOYSA-N
Synonym 4-aminoanisole-3-sulfonic acid, p-anisidine-3-sulfonic acid, benzenesulfonic acid, 2-amino-5-methoxy, 4-methoxy-2-sulfoaniline, 2-amino-5-methoxybenzenesulphonic acid, unii-hju535j932, 2-amino-5-methoxybenzene-1-sulfonic acid, 4-methoxyaniline-2-sulfonic acid, 4-anisidine-2-sulfonicacid, 2-amino-5-methoxy benzenesulfonic acid
PubChem CID 83260
IUPAC Name 2-amino-5-methoxybenzenesulfonic acid
SMILES COC1=CC(=C(C=C1)N)S(=O)(=O)O

Specifications

Quantity 25 g
Formula Weight 203.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material p-Anisidine-2-sulfonic Acid
EINECSNumber (3)-2039
RTECSNumber DB4950000
TSCA Yes