missing translation for 'onlineSavingsMsg'
Learn More

p-Anisaldehyde Dimethyl Acetal 97.0+%, TCI America™

Catalog No. A1247500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1247500ML 500 mL
A124725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1247500ML Supplier TCI America Supplier No. A1247500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2186-92-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00036507
InChI Key NNHYAHOTXLASEA-UHFFFAOYSA-N
Synonym 1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal
PubChem CID 75140
IUPAC Name 1-(dimethoxymethyl)-4-methoxybenzene
SMILES COC(OC)C1=CC=C(OC)C=C1

Specifications

Color Yellow
Boiling Point 253°C
Quantity 500 mL
Formula Weight 182.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material p-Anisaldehyde Dimethyl Acetal