accessibility menu, dialog, popup

UserName

CAS RN 2186-92-7

IUPAC Name: 1-(dimethoxymethyl)-4-methoxybenzene
Synonyms: 1-(dimethoxymethyl)-4-methoxybenzene
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: NNHYAHOTXLASEA-UHFFFAOYSA-N
SMILES: COC(OC)C1=CC=C(OC)C=C1
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)