missing translation for 'onlineSavingsMsg'
Learn More

p-Anisaldehyde, 98%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor AN122100ML

 View more versions of this product

Catalog No. 18612129


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

123-11-5
248°C
100 mL
Reagent
1
Poly Drum
0.98
Liquid

Chemical Identifiers

123-11-5
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.