missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Acetanisidide 98.0+%, TCI America™
Supplier: TCI America A0019100G
Specifications
| p-Acetanisidide | |
| 130°C | |
| C9H11NO2 | |
| 100 g | |
| XVAIDCNLVLTVFM-UHFFFAOYSA-N | |
| N-(4-methoxyphenyl)acetamide | |
| 5827 | |
| ≥98.0% (HPLC,N) |
| 51-66-1 | |
| White | |
| MFCD00014963 | |
| n-4-methoxyphenyl acetamide, 4'-methoxyacetanilide, p-methoxyacetanilide, p-acetanisidide, p-acetanisidine, methacetin, 4-methoxyacetanilide, aceto-p-anisidide, n-acetyl-p-anisidine, acetamide, n-4-methoxyphenyl | |
| CC(=O)NC1=CC=C(C=C1)OC | |
| 165.192 | |
| 165.19 | |
| Crystalline Powder |
Chemical Identifiers
| 51-66-1 | |
| 165.192 | |
| XVAIDCNLVLTVFM-UHFFFAOYSA-N | |
| 5827 | |
| CC(=O)NC1=CC=C(C=C1)OC |
| C9H11NO2 | |
| MFCD00014963 | |
| n-4-methoxyphenyl acetamide, 4'-methoxyacetanilide, p-methoxyacetanilide, p-acetanisidide, p-acetanisidine, methacetin, 4-methoxyacetanilide, aceto-p-anisidide, n-acetyl-p-anisidine, acetamide, n-4-methoxyphenyl | |
| N-(4-methoxyphenyl)acetamide |
Safety and Handling
EINECSNumber : (3)-0690
RTECSNumber : AE8290000
TSCA : Yes