accessibility menu, dialog, popup

UserName

CAS RN 51-66-1

H3CONHCH3O

CAS RN 51-66-1

IUPAC Name: N-(4-methoxyphenyl)acetamide
Synonyms: P-methoxyacetanilide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: XVAIDCNLVLTVFM-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(C)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results