missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxymatrine, 98%
$263.03 - $752.01
Chemical Identifiers
| CAS | 16837-52-8 |
|---|---|
| Molecular Weight (g/mol) | 264.37 |
| MDL Number | MFCD00210339 |
| InChI Key | XVPBINOPNYFXID-LHDUFFHYSA-N |
| IUPAC Name | (1R,2R,9S,17S)-6-oxo-7,13-diazatetracyclo[7.7.1.0²,â·.0¹³,¹â·]heptadecan-13-ium-13-olate |
| SMILES | [O-][N+]12CCC[C@H]3CN4[C@H](CCCC4=O)[C@@H](CCC1)[C@@H]23 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH37340MD
|
Thermo Scientific™
H37340MD |
250 mg |
Each for $263.03
|
|
|||||
|
AAH3734003
|
Thermo Scientific™
H3734003 |
1 g |
Each for $752.01
|
|
|||||
Chemical Identifiers
| 16837-52-8 | |
| MFCD00210339 | |
| (1R,2R,9S,17S)-6-oxo-7,13-diazatetracyclo[7.7.1.0²,â·.0¹³,¹â·]heptadecan-13-ium-13-olate |
| 264.37 | |
| XVPBINOPNYFXID-LHDUFFHYSA-N | |
| [O-][N+]12CCC[C@H]3CN4[C@H](CCCC4=O)[C@@H](CCC1)[C@@H]23 |
Specifications
| 16837-52-8 | |
| C{1}{5}H{2}{4}N{2}O{2} | |
| 250 mg | |
| 98% |
| 208° | |
| MFCD00210339 | |
| 264.37 | |
| Solid |
Safety and Handling
ShelfLife : 5 Years
EINECSNumber : 605-514-4
RTECSNumber : OQ1770000