missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxymatrine, 98%
Supplier: Thermo Scientific™ H3734003
Specifications
| 16837-52-8 | |
| C15H24N2O2 | |
| 1 g | |
| [O-][N+]12CCC[C@H]3CN4[C@H](CCCC4=O)[C@@H](CCC1)[C@@H]23 | |
| 264.37 | |
| 98% |
| 208° | |
| MFCD00210339 | |
| XVPBINOPNYFXID-LHDUFFHYSA-N | |
| (1R,2R,9S,17S)-6-oxo-7,13-diazatetracyclo[7.7.1.0²,â·.0¹³,¹â·]heptadecan-13-ium-13-olate | |
| 264.37 | |
| Solid |
Chemical Identifiers
| 16837-52-8 | |
| MFCD00210339 | |
| (1R,2R,9S,17S)-6-oxo-7,13-diazatetracyclo[7.7.1.0²,â·.0¹³,¹â·]heptadecan-13-ium-13-olate |
| 264.37 | |
| XVPBINOPNYFXID-LHDUFFHYSA-N | |
| [O-][N+]12CCC[C@H]3CN4[C@H](CCCC4=O)[C@@H](CCC1)[C@@H]23 |
Safety and Handling
ShelfLife : 5 Years
EINECSNumber : 605-514-4
RTECSNumber : OQ1770000