CAS RN 16837-52-8
CAS RN 16837-52-8
IUPAC Name:
(1R,2R,9S,17S)-6-oxo-7,13-diazatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadecan-13-ium-13-olate
Synonyms:
oxymatrine|ammothamnine
Molecular Weight (g/mol):
264.37
Molecular Formula:
C15H24N2O2
InChi Key:
XVPBINOPNYFXID-LHDUFFHYSA-N
SMILES:
[O-][N+]12CCC[C@H]3CN4[C@H](CCCC4=O)[C@@H](CCC1)[C@@H]23