missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxolinic acid, 98%
Targets gram-negative bacteria | CAS: 14698-29-4 | C13H11NO5 | 261.23 g/mol
$300.10 - $1,271.01
Chemical Identifiers
| CAS | 14698-29-4 |
|---|---|
| Molecular Formula | C13H11NO5 |
| Molecular Weight (g/mol) | 261.23 |
| MDL Number | MFCD00056775 |
| InChI Key | KYGZCKSPAKDVKC-UHFFFAOYSA-N |
| Synonym | 5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid |
| PubChem CID | 4628 |
| ChEBI | CHEBI:138856 |
| IUPAC Name | 5-ethyl-8-oxo-2H,5H,8H-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid |
| SMILES | CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ6663706
|
Thermo Scientific Chemicals
J6663706 |
5 g |
N/A
|
|
|||||
|
AAJ6663714
|
Thermo Scientific Chemicals
J6663714 |
25 g |
N/A
|
|
|||||
Description
Targets Gram-negative bacteria
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14698-29-4 | |
| 261.23 | |
| KYGZCKSPAKDVKC-UHFFFAOYSA-N | |
| 4628 | |
| 5-ethyl-8-oxo-2H,5H,8H-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid |
| C13H11NO5 | |
| MFCD00056775 | |
| 5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid | |
| CHEBI:138856 | |
| CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12 |
Specifications
| Oxolinic Acid | |
| 14698-29-4 | |
| White | |
| >110°C (230°F) | |
| C13H11NO5 | |
| 473°C | |
| 14,6965 | |
| Soluble in 0.5N NaOH with warming | |
| CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12 | |
| 261.23 | |
| CHEBI:138856 | |
| 98% |
| Powder | |
| 314°C to 316°C (decomposition) | |
| 5 g | |
| Light sensitive | |
| MFCD00056775 | |
| 620635 | |
| 5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid | |
| KYGZCKSPAKDVKC-UHFFFAOYSA-N | |
| 5-ethyl-8-oxo-2H,5H,8H-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid | |
| 4628 | |
| 261.23 | |
| Oxolinic acid |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 238-750-8
RTECSNumber : JI5075000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only