Learn More
Oxolinic acid, 98%
Targets gram-negative bacteria | CAS: 14698-29-4 | C13H11NO5 | 261.23 g/mol
Supplier: Thermo Scientific Chemicals J6663714
Description
Targets Gram-negative bacteria
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Oxolinic Acid | |
| Powder | |
| 314°C to 316°C (decomposition) | |
| 25 g | |
| Light sensitive | |
| MFCD00056775 | |
| 620635 | |
| Soluble in 0.5N NaOH with warming | |
| CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12 | |
| 261.23 | |
| CHEBI:138856 | |
| 98% |
| Oxolinic acid | |
| 14698-29-4 | |
| Plastic Bottle | |
| >110°C (230°F) | |
| C13H11NO5 | |
| 473°C | |
| 5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid | |
| KYGZCKSPAKDVKC-UHFFFAOYSA-N | |
| 5-ethyl-8-oxo-2H,5H,8H-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid | |
| 4628 | |
| 261.23 |
Chemical Identifiers
| 14698-29-4 | |
| 261.23 | |
| KYGZCKSPAKDVKC-UHFFFAOYSA-N | |
| 4628 | |
| 5-ethyl-8-oxo-2H,5H,8H-[1,3]dioxolo[4,5-g]quinoline-7-carboxylic acid |
| C13H11NO5 | |
| MFCD00056775 | |
| 5,8-Dihydro-5-ethyl-8-oxo-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid | |
| CHEBI:138856 | |
| CCN1C=C(C(O)=O)C(=O)C2=CC3=C(OCO3)C=C12 |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 238-750-8
RTECSNumber : JI5075000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only