missing translation for 'onlineSavingsMsg'
Learn More

Oxibendazole, Thermo Scientific™

Catalog No. AC468810050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468810050 5 g
AC468810010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468810050 Supplier Thermo Scientific Chemicals Supplier No. 468810050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20559-55-1
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number 00133728
InChI Key RAOCRURYZCVHMG-UHFFFAOYSA-N
IUPAC Name methyl N-(6-propoxy-1H-1,3-benzodiazol-2-yl)carbamate
SMILES CCCOC1=CC=C2N=C(NC(=O)OC)NC2=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color White to Beige
Merck Index 13-7005
Solubility Insoluble in water
Quantity 5 g
Chemical Name or Material Oxibendazole
EINECSNumber 243-877-7
RTECSNumber FD0835000
Recommended Storage Normal conditions