missing translation for 'onlineSavingsMsg'
Learn More

Oxfendazole, Thermo Scientific™

Catalog No. AC469210050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469210050 5 g
AC469210010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC469210050 Supplier Thermo Scientific Chemicals Supplier No. 469210050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53716-50-0
Molecular Formula C15H13N3O3S
Molecular Weight (g/mol) 315.35
MDL Number 00801063
InChI Key BEZZFPOZAYTVHN-UHFFFAOYNA-N
IUPAC Name methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder/Crystals
Color White to Light Yellow to Beige
Merck Index 13-7004
Quantity 5 g
Chemical Name or Material Oxfendazole
Hazard Category Exclamation mark
EINECSNumber 258-714-5
RTECSNumber FD0808000
Recommended Storage Normal conditions