accessibility menu, dialog, popup

UserName

CAS RN 53716-50-0

IUPAC Name: methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate
Synonyms: oxfendazole
Molecular Weight (g/mol): 315.35
Molecular Formula: C15H13N3O3S
InChi Key: BEZZFPOZAYTVHN-UHFFFAOYNA-N
SMILES: COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results