missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxfendazole, Thermo Scientific™
Specifications
Specifications
| Packaging | Glass Bottle |
| Certificate of Analysis | Available |
| Physical Form | Powder/Crystals |
| Color | White to Light Yellow to Beige |
| Merck Index | 13-7004 |
| Quantity | 1 g |
| Chemical Name or Material | Oxfendazole |
Safety and Handling
| Hazard Category | Exclamation mark |
| EINECSNumber | 258-714-5 |
| RTECSNumber | FD0808000 |
| Recommended Storage | Normal conditions |