missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxamic Acid 97.0+%, TCI America™
$59.69 - $175.20
Chemical Identifiers
| CAS | 471-47-6 |
|---|---|
| Molecular Formula | C2H3NO3 |
| Molecular Weight (g/mol) | 89.05 |
| MDL Number | MFCD00008006 |
| InChI Key | SOWBFZRMHSNYGE-UHFFFAOYSA-N |
| Synonym | 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent |
| PubChem CID | 974 |
| ChEBI | CHEBI:18058 |
| IUPAC Name | oxamic acid |
| SMILES | C(=O)(C(=O)O)N |
Chemical Identifiers
| 471-47-6 | |
| 89.05 | |
| SOWBFZRMHSNYGE-UHFFFAOYSA-N | |
| 974 | |
| oxamic acid |
| C2H3NO3 | |
| MFCD00008006 | |
| 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent | |
| CHEBI:18058 | |
| C(=O)(C(=O)O)N |
Specifications
| 471-47-6 | |
| White | |
| MFCD00008006 | |
| 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent | |
| C(=O)(C(=O)O)N | |
| 89.05 | |
| CHEBI:18058 | |
| ≥97.0% (T,N) | |
| Oxamic Acid |
| 217°C | |
| C2H3NO3 | |
| 5 g | |
| SOWBFZRMHSNYGE-UHFFFAOYSA-N | |
| oxamic acid | |
| 974 | |
| 89.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes