accessibility menu, dialog, popup

UserName

H2NOOHO

CAS RN 471-47-6

IUPAC Name: carbamoylformic acid
Synonyms: oxamic acid oxalic monoamide oxamate, (aminocarbonyl)- (aminocarbonyl)-oxamate
Molecular Weight (g/mol): 89.05
Molecular Formula: C2H3NO3
InChi Key: SOWBFZRMHSNYGE-UHFFFAOYSA-N
SMILES: NC(=O)C(O)=O
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)