missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Oxamic Acid | |
| 217°C | |
| C2H3NO3 | |
| 25 g | |
| SOWBFZRMHSNYGE-UHFFFAOYSA-N | |
| oxamic acid | |
| 974 | |
| 89.05 | |
| Crystalline Powder |
| 471-47-6 | |
| White | |
| MFCD00008006 | |
| 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent | |
| C(=O)(C(=O)O)N | |
| 89.05 | |
| CHEBI:18058 | |
| ≥97.0% (T,N) |
Chemical Identifiers
| 471-47-6 | |
| 89.05 | |
| SOWBFZRMHSNYGE-UHFFFAOYSA-N | |
| 974 | |
| oxamic acid |
| C2H3NO3 | |
| MFCD00008006 | |
| 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent | |
| CHEBI:18058 | |
| C(=O)(C(=O)O)N |
Safety and Handling
TSCA : Yes