missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid, 10% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 54701
Specifications
| Oxalic Acid | |
| 89.0,8.91 | |
| 93.0,9.28 | |
| oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
| C(=O)(C(=O)O)O.O.O | |
| oxalic acid;dihydrate | |
| 61373 | |
| Laboratory | |
| Natural Poly Bottle |
| 7732-18-5,6153-56-6 | |
| Colorless | |
| C2H6O6 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| 10% (w/v) | |
| Mixture | |
| 10% w/v | |
| 4 L | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm | |
| oxalic acid;dihydrate |
| C2H6O6 | |
| GEVPUGOOGXGPIO-UHFFFAOYSA-N | |
| 61373 | |
| C(=O)(C(=O)O)O.O.O |