missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, 10% (w/v), Ricca Chemical

Catalog No. p-3252019
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
547016 500 mL Natural Poly Bottle
547032 1 L Natural Poly Bottle
54701 4 L Natural Poly Bottle
R547000020F 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 547016 Supplier Ricca Chemical Company Supplier No. 547016
Only null left

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) Mixture
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

CAS Min % 89.0, 8.91
Color Colorless
CAS Max % 93.0, 9.28
Concentration 10% (w/v)
Percent Purity 10% w/v
Grade Laboratory
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Oxalic Acid