missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalic Acid, 0.100 Normal (N/10), Reagents
Oxalic Acid, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CO1000004A
Specifications
| Oxalic Acid, 0.100 Normal (N/10) | |
| 7732-18-5,6153-56-6 | |
| Colorless | |
| Ethanedioic Acid | |
| O | |
| 18.02 | |
| 4 L | |
| Liquid |
| <2 | |
| 100°C | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| Poly Bottle | |
| 1.0025 |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Ethanedioic Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |