missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid, 0.100 Normal (N/10), Reagents

Catalog No. p-200021634
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334569 1 L
01334568 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01334569 Supplier Labchem Inc Supplier No. CO1000001C
Only null left

Oxalic Acid, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 6153-56-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanedioic Acid
IUPAC Name water
SMILES O

Specifications

pH <2
Boiling Point 100°C
Color Colorless
Grade Reagent
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.0025
Chemical Name or Material Oxalic Acid, 0.100 Normal (N/10)