missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organometallic Oil Standard, 12-Element, SPEX CertiPrep™
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Ag | 500μg/g |
| Al | 500μg/g |
| Cr | 500μg/g |
| Cu | 500μg/g |
| Fe | 500μg/g |
| Mg | 500μg/g |
| Na | 500μg/g |
| Ni | 500μg/g |
| Pb | 500μg/g |
| Si | 500μg/g |
| Sn | 500μg/g |
| Ti | 500μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | 12 Element Oil Standard |
| Packaging | Triple-leached LDPE bottle |
| Certificate of Analysis | With Certificate of Analysis. |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Base Oil 75 |
| Industry Type | Petrochemical Testing |
| Quantity | 50 g |
| Grade | Oil |
| Show More |
Product intended for laboratory use only