missing translation for 'onlineSavingsMsg'
Learn More

Organometallic Oil Standard, 12-Element, SPEX CertiPrep™

Catalog No. S12100Z
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S12100Z 50 g
S12100Y 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S12100Z Supplier SPEX CertiPrep Group LLC Supplier No. S12100Z
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag100μg/g
Al100μg/g
Cr100μg/g
Cu100μg/g
Fe100μg/g
Mg100μg/g
Na100μg/g
Ni100μg/g
Pb100μg/g
Si100μg/g
Sn100μg/g
Ti100μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material 12 Element Oil Standard
Packaging Triple-leached LDPE bottle
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Base Oil 75
Industry Type Petrochemical Testing
Quantity 50 g
Grade Oil
Concentration or Composition (by Analyte or Components) Each component at 100μg/g (S12-100), 300μg/g (S12-300), 500μg/g (S12-500), or 900μg/g (S12-900) in Base Oil 75:
Aluminum
Chromium
Copper
Iron
Lead
Magnesium
Nickel
Silicon
Silver
Sodium
Tin
Titanium
Show More Show Less

Product intended for laboratory use only