missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Standard, 5000 ppm C, Ricca Chemical
$152.84 - $152.84
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C8H5KO4 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | IWZKICVEHNUQTL-UHFFFAOYSA-M |
| PubChem CID | 23676735 |
| IUPAC Name | potassium;2-carboxybenzoate |
| SMILES | C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R1849500500
|
Ricca Chemical Company
R1849500500C |
500 mL | Amber Glass Bottle |
Each for $152.84
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| 23676735 | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
| C8H5KO4 | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate |
Specifications
| Amber Glass Bottle | |
| Odorless | |
| Liquid | |
| Colorless | |
| Infinitely soluble in water | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| Water | |
| Mixture | |
| 23676735 | |
| Organic Carbon Standard, 1mL = 5mg C, 5000ppm C for Total Organic Carbon (TOC) |
| 7732-18-5,877-24-7,7732-18-5 | |
| 5, 000 ± 10ppm | |
| C8H5KO4 | |
| 100°C | |
| Standard | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
| potassium;2-carboxybenzoate | |
| 500 mL | |
| Laboratory |