missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 50 ppm C, Ricca Chemical

Catalog No. R1844500250
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1844500250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1844500250 Supplier Ricca Chemical Company Supplier No. R1844500250C
Only null left

Organic Carbon Standard

Chemical Identifiers

CAS 877-24-7 , 7664-38-2 , 7732-18-5
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Glass Amber Bottle
CAS Min % 99.59, 0.4, 0.01
CAS Max % 99.59, 0.4, 0.01
Density 1
Physical Form Liquid
Color Colorless
pH <2
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
Quantity 250 mL
Chemical Name or Material Organic Carbon Standard, 50 ppm C
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient