missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 300 ppm C, Ricca Chemical

Supplier:  Ricca Chemical Company R18463001C

Catalog No. R18463001C


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Organic Carbon Standard
Amber Glass Bottle
C8H5KO4
0.0°C
Water Analyzer
Calibration Solution
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
1 L
Laboratory
300 ppm C
877-24-7 , 7664-38-2 , 7732-18-5
Colorless
100°C
Miscible with water
TOC Standard
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
23676735

Chemical Identifiers

877-24-7
204.222
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 months

SDS
Documents

Documents

Promotions

Promotions