missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 200 ppm C, Ricca Chemical

Catalog No. R1846200250
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R1846200250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1846200250 Supplier Ricca Chemical Company Supplier No. R1846200250C

Chemical Identifiers

CAS 877-24-7 , 7664-38-2 , 7732-18-5
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Physical Form Liquid
Color Colorless
pH < 2
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Miscible with water
Solution Type Calibration Solution
Type TOC Standard
Quantity 250 mL
Grade Laboratory
Chemical Name or Material Organic Carbon Standard, 200 ppm C
Show More Show Less
ShelfLife 12 months