missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 2000 ppm C, Ricca Chemical

Catalog No. R18480001C
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R18480001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R18480001C Supplier Ricca Chemical Company Supplier No. R18480001C
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2 , 877-24-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 23676735
IUPAC Name water
SMILES O

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Assay Percent Range 2, 000 ± 5ppm
Color Colorless
pH <2
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Solubility Infinitely soluble in water
Solution Type Calibration Standard
Type Standard
Solvent or Matrix Water
Quantity 1 L
Grade Laboratory
Chemical Name or Material Organic Carbon Standard, 1mL = 2mg C, 2000ppm C for Total Organic Carbon (TOC)
Show More Show Less