missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 100 ppm C, Ricca Chemical

Catalog No. R1846000250
Change view
Click to view available options
Quantity:
250 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R1846000250 250 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R1846000250 Supplier Ricca Chemical Company Supplier No. R1846000250C
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2 , 877-24-7
Molecular Formula C8H5KO4
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Amber Glass Bottle
CAS Min % 99.58, 0.4, 0.02
CAS Max % 99.58, 0.4, 0.02
Density 1.001
Physical Form Liquid
Color Colorless
pH < 2
Melting Point 0°C
Boiling Point 100°C
Solubility Miscible with water
Type Standard
Molecular Weight (g/mol) 204.222
Quantity 250 mL
Grade Laboratory
Chemical Name or Material Organic Carbon Standard, 100 ppm C
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient