missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Standard, 50 ppm C, Ricca Chemical
$235.63 - $324.19
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C8H5KO4 |
| Molecular Weight (g/mol) | Mixture |
| InChI Key | IWZKICVEHNUQTL-UHFFFAOYSA-M |
| PubChem CID | 23676735 |
| IUPAC Name | potassium;2-carboxybenzoate |
| SMILES | C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R18445001C
|
Ricca Chemical Company
R18445001C |
1 L | Amber Glass Bottle |
Each for $235.63
|
|
||||
|
R18445004C
|
Ricca Chemical Company
R18445004C |
4 L | Amber Glass Bottle |
Each for $324.19
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| 23676735 | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] |
| C8H5KO4 | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| potassium;2-carboxybenzoate |
Specifications
| Amber Glass Bottle | |
| Odorless | |
| Liquid | |
| Colorless to Yellow | |
| Infinitely soluble in water | |
| IWZKICVEHNUQTL-UHFFFAOYSA-M | |
| Water | |
| Mixture | |
| 23676735 | |
| Organic Carbon Standard, 1mL = 0.050mg C, 50ppm C for Total Organic Carbon (TOC) |
| 7732-18-5,7664-38-2,877-24-7 | |
| 50.0 ± 0.5ppm | |
| C8H5KO4 | |
| 100°C | |
| Standard | |
| C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+] | |
| potassium;2-carboxybenzoate | |
| 1 L | |
| Laboratory |