missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Organic Carbon Standard, 50 ppm C, Ricca Chemical
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Odor | Odorless |
| Assay Percent Range | 50.0 ± 0.5ppm |
| Physical Form | Liquid |
| Color | Colorless to Yellow |
| Boiling Point | 100°C |
| Solubility | Infinitely soluble in water |
| Type | Standard |
| Solvent or Matrix | Water |
| Quantity | 4 L |
| Show More |