missing translation for 'onlineSavingsMsg'
Learn More

Organic Carbon Standard, 50 ppm C, Ricca Chemical

Catalog No. R18445001C
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R18445001C 1 L Amber Glass Bottle
R18445004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R18445001C Supplier Ricca Chemical Company Supplier No. R18445001C
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7664-38-2 , 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) Mixture
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Packaging Amber Glass Bottle
Odor Odorless
Assay Percent Range 50.0 ± 0.5ppm
Physical Form Liquid
Color Colorless to Yellow
Boiling Point 100°C
Solubility Infinitely soluble in water
Type Standard
Solvent or Matrix Water
Quantity 1 L
Grade Laboratory
Chemical Name or Material Organic Carbon Standard, 1mL = 0.050mg C, 50ppm C for Total Organic Carbon (TOC)
Show More Show Less